Anti-IL-2 Goat Polyclonal Antibody

Supplier: Peprotech
500-P22G-50UG 500-P22G-100UG 500-P22G-1MG
10775-122EA 565.59 CAD
10775-122 10775-124 10775-126
Anti-IL-2 Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human IL-2. Anti­Human IL-2­specific antibody was purified by affinity chromatography employing an immobilized Human IL-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR