Anti-FGF-10 Goat Polyclonal Antibody

Supplier: Peprotech
500-P151G-50UG 500-P151G-100UG 500-P151G-1MG
10771-440EA 560.17 CAD
10771-440 10771-442 10771-444
Anti-FGF-10 Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human FGF-10. Anti­Human FGF-10­specific antibody was purified by affinity chromatography employing an immobilized Human FGF-10 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: FGF-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR