Anti-Eotaxin-3 (CCL26) Goat Polyclonal Antibody

Supplier: Peprotech
500-P156G-50UG 500-P156G-100UG 500-P156G-1MG
10776-450EA 536.88 CAD
10776-450 10776-452 10776-454
Anti-Eotaxin-3 (CCL26) Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human Eotaxin-3 (CCL26). Anti­Human Eotaxin-3 (CCL26)­specific antibody was purified by affinity chromatography employing an immobilized Human Eotaxin-3 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Eotaxin-3 (CCL26)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR