Anti-EMAP-II Goat Polyclonal Antibody

Supplier: Peprotech
500-P172G-50UG 500-P172G-100UG 500-P172G-1MG
10776-586EA 536.88 CAD
10776-586 10776-588 10776-590
Anti-EMAP-II Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human EMAP-II. Anti­Human EMAP-II­specific antibody was purified by affinity chromatography employing an immobilized Human EMAP-II matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: EMAP-II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR