Anti-BAFF Goat Polyclonal Antibody

Supplier: Peprotech
500-P163G-50UG 500-P163G-100UG 500-P163G-1MG
10776-510EA 536.88 CAD
10776-510 10776-512 10776-514
Anti-BAFF Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human BAFF. Anti­Human BAFF­specific antibody was purified by affinity chromatography employing an immobilized Human BAFF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: BAFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR