Anti-TPO Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P49BT-25UG 500-P49BT-50UG
76077-366EA 8330.9 CAD
76077-366 76081-518 76081-520
Anti-TPO Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human TPO. Anti­Human TPO­specific antibody was purified by affinity chromatography employing an immobilized Human TPO matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: TPO
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR