Anti-NT-3 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P82BT-25UG 500-P82BT-50UG
76077-374EA 536.88 CAD
76077-374 76077-376 76077-378
Anti-NT-3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human NT-3. Anti­Human NT-3­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: NT-3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR