Anti-TPO Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P49-50UG 500-P49-100UG 500-P49-1MG
76077-362EA 536.88 CAD
76077-362 76077-364 76081-516
Anti-TPO Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human TPO. Anti­Human TPO­specific antibody was purified by affinity chromatography employing an immobilized Human TPO matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: TPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR