Anti-NT-3 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P82-50UG 500-P82-100UG 500-P82-1MG
76077-368EA 536.88 CAD
76077-368 76077-370 76077-372
Anti-NT-3 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human NT-3. Anti­Human NT-3­specific antibody was purified by affinity chromatography employing an immobilized Human NT-3 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: NT-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR