Anti-BD-5 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P323-50UG 500-P323-100UG 500-P323-1MG
76077-380EA 536.88 CAD
76077-380 76077-382 76077-384
Anti-BD-5 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human BD-5. Anti­Human BD-5­specific antibody was purified by affinity chromatography employing an immobilized Human BD-5 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: BD-5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR