Cole-Parmer® Stuart Digital Melting Point Apparatus, Antylia Scientific
Supplier: Antyila Scientific
03011-40 03011-45 03011-49
76269-138EA 3285.36 CAD
76269-138 76269-140 76269-142
Cole-Parmer® Stuart Digital Melting Point Apparatus, Antylia Scientific
Melting Point Apparatus
No more reading glass thermometers.
  • Select, measure, and display the temperature digitally, ensuring accuracy and repeatability
  • Easily view the melt via magnifying lens with built-in illumination
  • Be more productive—test multiple samples simultaneously
  • Save time by setting a plateau temperature for initial warm-up
Designed for maximum safety and ease of use. Select, measure, and display the temperature digitally, ensuring accuracy and avoiding the need for a glass thermometer. Test two samples simultaneously, viewing the melt via a magnifying lens withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Memory 7 results per tube