Cole-Parmer® MP-100 Series Stuart Analog Melting Point Apparatus, Antylia Scientific
Supplier: Antyila Scientific
03013-60
76268-912EA 2784.04 CAD
76268-912
Cole-Parmer® MP-100 Series Stuart Analog Melting Point Apparatus, Antylia Scientific
Melting Point Apparatus
Integral LED illumination and magnifying lens make viewing easy.
  • Simple to use—place sample in capillary tube, set heating rate, and watch for melt
  • Test and view three samples at the same time
  • Heat up to 20°C/min close to melt temperature then set to slower heating rate of 1 to 10 °C/min
  • Read temperature mercury-free blue-spirit-filled thermometer
Take melting point measurements of up to three samples at once for greater efficiency. Easy-to-follow instructions are printed directly on the unit. Place the sample in a capillary tube and insert it into the aluminum block. Manually set the heating rate between 1 and 20 °C per minute, up to a maximum of 250 °C. Lowering the heating rate as you near the expected melting point temperature will increase the accuracy of the reading. Observe the sample through the magnifying lens until the melt occurs and read the melt temperature from the thermometer.
Certifications: CE Approved.
Accessories information: Accessory glass capillary tubes are constructed of soda glass and easily sealed in a Bunsen flame. Supplied in a durable tube holder for added protection. Ideal for your melting point apparatus with 100 mm length, 1.9 mm Ext.Ø., 1.3 mm Int.Ø., and 0.3 mm wall thickness.
Ordering information: Apparatus is supplied with open-ended capillary tubes. One end should be sealed with a Bunsen burner; or purchase optional capillary tubes separately that are already sealed. Order the optional brass cooling plug to speed up cooling when multiple runs are needed.
Supplied with pack of 100 capillary tubes (open at both ends) and power cord with US plug.