Quantum Levitation DIY Maglev Kits
Supplier: QUANTUM EXPERIENCE LTD
301-111 302-112 302-122
470306-214EA 1014.69 CAD
470306-214 470306-216 470306-218
Quantum Levitation DIY Maglev Kits
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Build your own Quantum Levitation maglev track!
  • Easy to use
  • Highly durable
Use super-strong NdFeB magnets to construct a flexible linear magnetic track. Experience frictionless motion in both levitation and suspension. Excellent for science projects and science classes. Adult supervision is required during construction and operation.
Ordering information: The Pro Kit comes with Small / Medium Quantum Levitator; Plastic tweezers; 150 / 300 NdFeB block magnets (10×10×5 mm3); Stainless steel track 40/80 cm; and User manual.