Quantum Levitation MiniMaglev Kit
Supplier: QUANTUM EXPERIENCE LTD
304-112 305-112-122
470306-206EA 2848.99 CAD
470306-206 470306-208
Quantum Levitation MiniMaglev Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Experience Quantum Levitation with this portable, light weight and easy to use maglev track.
  • Easy to use
  • Highly durable
The track uses super-strong NdFeB magnets in a unique double rings design to achieve maximal levitation height. Demonstrate frictionless motion in both levitation and suspension.
The handheld magnetic device (Starter Kit) can be passed around for a captivating hands-on experience.
Excellent for science demonstrations in schools and universities, undergraduate and high-school experiments in quantum and classical physics (energyMore Product Information