Quantum Enhanced Levitators
Supplier: QUANTUM EXPERIENCE LTD
12-2 12-3 12-4
470306-194EA 1184.99 CAD
470306-194 470306-196 470306-198
Quantum Enhanced Levitators
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Quantum Levitators are intended for science demonstrations and visual display.
  • Easy to use
  • Highly durable
The levitators are provided with a 6 months full no-questions-asked warranty. The Quantum Enhanced Levitators, having extra superconducting material, are able to carry light weight (<200 g) when used in conjunction with a suitable magnetic setup or track. The X strong Enhanced Levitators can carry weight of 500 g.
Order Now
SPECIFICATIONS
Environmentally Preferable