Quantum Standard Levitators
Supplier: QUANTUM EXPERIENCE LTD
11-1 11-2 11-3 73-459
470306-188EA 607.29 CAD
470306-188 470306-190 470306-192 470306-212
Quantum Standard Levitators
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Quantum Levitators are intended for science demonstrations.
  • Easy to use
  • Highly durable
The levitators are provided with 6 months full no-questions-asked warranty. The Quantum Levitators are able to carry only their own weight plus liquid nitrogen.
The Starter Kit is the most fundamental, entry level for classroom demonstration of Quantum Levitation and Flux Pinning. Perfect for high-school students as well as university undergraduates; the kit is easy to use, highly durable and portable. Use the rectangle magnetic setup to witness the Quantum Locking phenomena and the round magnetic setup to demonstrate frictionless motion.
When used with the new resistance thermometer equipped Ed levitator, the handheld magnetic device serves as a unique experimentation platform where quantum phenomena such as the Meissner effect and flux pinning can be investigated.