Mouse Recombinant IL-22 (from E. coli)
Supplier: Peprotech
AF-210-22-2UG AF-210-22-10UG AF-210-22-50UG AF-210-22-100UG AF-210-22-250UG AF-210-22-500UG AF-210-22-1MG
76077-488EA 266.99 CAD
76077-488 76077-490 76077-492 76077-494 76077-496 76077-498 76077-500
Mouse Recombinant IL-22 (from E. coli)
Proteins and Peptides
  • Animal-free
IL-22 is a member of the IL-10 family of regulatory cytokines, which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Members of this family share partial homology in their amino acid sequences, but they are dissimilar in their biological functions. Produced by T lymphocytes, IL-22 inhibits IL-4 production by Th2 cells, and induces acute phase reactants in the liver and pancreas. IL-22 signals through a receptor system consisting of IL-10Rβ/CRF2-4 and IL-22R, both of which are members of the class II cytokine-receptor family. Recombinant Murine IL-22 is a 33.4 kDa, non-disulfide-linked, homodimeric protein consisting of two 147 amino acid polypeptide chains.