Human Recombinant TREM-1 (from CHO Cells)
Supplier: Peprotech
310-36-10UG 310-36-50UG 310-36-100UG 310-36-250UG 310-36-500UG 310-36-1MG
75999-604EA 209.84 CAD
75999-604 75999-606 75999-608 75999-610 75999-612 75999-614
Human Recombinant TREM-1 (from CHO Cells)
Proteins and Peptides
Triggering receptor expressed on myeloid cells-1 (TREM-1) is a type 1 transmembrane glycoprotein belonging to the immunoglobulin superfamily of cell surface receptors that is constitutively expressed on the surface of monocytes, neutrophils and macrophages. The first identified member of the TREM family of receptors, TREM-1 is a proponent of amplified inflammatory responses triggered by bacterial and fungal infection, and functions in association with the signal transduction adaptor molecule DAP12. TREM-1 activation induces the sustained secretion of proinflammatory chemokines (IL-8 and MCP-1) and cytokines (TNF-α, IL-1β, and IL-6), the respiratory burst and degranulation of neutrophils, the upregulation of adhesion molecules involved in leukocyte extravasation, and tissue degradation. Involvement and increased expression of TREM-1, or soluble TREM-1, in infectious, as well as some non-infectious, inflammatory conditions may highlight the protein’s potential as a possible diagnostic marker in cases of severe inflammation, such as myocardial dysfunction related to severe sepsis and spontaneous preterm labor. TREM-1 involvement in inflammatory response has also been noted as a predictor of both progression and aggression in certain cancers. PeproTech’s