Human Recombinant sCD28 (from CHO Cells)
Supplier: Peprotech
310-34-20UG 310-34-100UG 310-34-250UG 310-34-500UG 310-34-1MG
75999-584EA 221.22 CAD
75999-584 75999-586 75999-588 75999-590 75999-592
Human Recombinant sCD28 (from CHO Cells)
Proteins and Peptides
CTLA-4 and CD28 are receptors of the immunoglobulin superfamily that are expressed, along with the transmembrane glycoproteins B7-1 and B7-2, by antigen-presenting cells, and with these ligands constitute crucial co-stimulatory pathways for T and B cell regulatory responses. It is through engagement with CD28 and CTLA-4 that the B7 family ligands B7-1 and B7-2 play principal roles in immunity by activating immune response and maintaining immune tolerance. Co-stimulatory signals generated by B7-1 and B7-2 interactions with CD28 serve to stimulate T cell activation and prevent anergy through the amplification of T cell receptor (TCR) signaling. In contrast, interactions of the ligands with CTLA-4 serves to maintain T cell homeostasis and self-tolerance through the disruption of stimulatory signaling from B7 isoform-bound CD28 complexes, and by inducing powerful inhibitory signals in T cells. CTLA-4, like B7-1, is only poorly expressed on resting dendritic cells; therefore, up-regulation of their interaction and resultant amplification and regulation of T cell activity at peripheral inflammation sites is considerably delayed upon immune activation. Conversely, B7-2 and CD28 are constitutively expressed by resting hematopoietic and T cells, respectively, and as a result are able to rapidly induce up-regulation upon immune activation, making them critical to the early co-stimulatory signaling of immune response. Unlike B7-1 and B7-2, the ligands PD-L1 (or B7-H1) and B7-H2, which also belong to the B7 family, have not been shown to influence immunity through interaction with CTLA-4. B7-H2 has been shown to have restricted interaction with CD28. The difference in expression of B7-1, B7-2 and B7-H2 may enable temporally and spatially-specific regulation of T cell response through non-competitive CD28 interaction. PeproTech’s