Human Recombinant Mesothelin (from CHO Cells)
Supplier: Peprotech
100-63-10UG 100-63-50UG 100-63-100UG 100-63-250UG 100-63-500UG 100-63-1MG
75999-390EA 209.84 CAD
75999-390 75999-392 75999-394 75999-396 75999-398 75999-400
Human Recombinant Mesothelin (from CHO Cells)
Proteins and Peptides
Originally identified as a differentiation antigen of mesotheliomas, ovarian cystadenocarcinomas, and pancreatic adenocarcinomas, mesothelin is a glycosylphosphatidylinositol (GPI)-anchored, cell-surface glycoprotein predominantly secreted by cells of the mesothelium. Although mesothelin is expressed at restricted levels by normal mesothelial cells of the pleural, pericardial, and peritoneal membranes, aberrant expression has been documented in the aforementioned cancers, as well as in endometrioid uterine adenocarcinomas and squamous cell carcinomas of the esophagus, stomach, lung, and cervix. Proteolytic cleavage of mesothelin yields a soluble, polypeptide fragment-designated megakaryocyte-potentiating factor (MPF) based on its ability to stimulate megakaryocyte colony-forming activity of murine IL-3 in murine bone marrow cell cultures. Originally isolated from the HPC-Y5 pancreatic cell line, MPF has been suggested to play a role in the proliferation and differentiation of megakaryoctyes, and the regulation of resultant platelet production. While the biological functions of both mesothelin and MPF remain speculative, high levels of expression in cancerous tissues, compared to limited distribution in normal tissues, strongly suggests their involvement in tumorigenesis. Both have been demonstrated to promote tumor cell proliferation, migration, anchorage-independent growth, and tumor progression, demonstrating their involvement in heterotypic cell adhesion and the metastatic spread of cancer. PeproTech’s