Human Recombinant Leptin (from E. coli)
Supplier: Peprotech
AF-300-27-200UG AF-300-27-1MG AF-300-27-5MG
75999-758EA 250.97 CAD
75999-758 75999-760 75999-762
Human Recombinant Leptin (from E. coli)
Proteins and Peptides
  • Animal-free
Encoded by the ob (obese) gene, Leptin is an adipose-derived cytokine that suppresses appetite and increases thermogenesis. Leptin exerts its anorectic effect via signaling through a hypothalamic receptor termed OB-R. Leptin has been shown to reduce body weight, food consumption, and plasma glucose levels in various Recombinant Human Leptin is a 16.0 kDa protein containing 147 amino acid residues.