Human Recombinant IL-37 (from E. coli)
Supplier: Peprotech
200-39-5UG 200-39-25UG 200-39-100UG 200-39-250UG 200-39-500UG 200-39-1MG
76077-344EA 209.84 CAD
76077-344 76077-346 76077-348 76077-350 76077-352 76077-354
Human Recombinant IL-37 (from E. coli)
Proteins and Peptides
The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-37 (IL-1F7), which acts as a modulator of the immune response. Reduction of IL-37 synthesis in PBMCs leads to increased production of pro-inflammatory cytokines including IL-1 alpha, IL-1 beta, IL-6 and TNF- alpha. The role of IL-37 as an inhibitor of the innate inflammatory response is also corroborated by the observation that it is highly expressed in synovial tissue from patients with rheumatoid arthritis. Full length IL-37 resides primarily in the cytoplasm, but after activation through cleavage by CASP1, it can translocate to the nucleus where it exerts its activity by direct interaction with SMAD3. Recombinant Human IL-37 is a 19.4 kDa protein consisting of 174 amino acids, corresponding to the mature activated form of IL-37.