Human Recombinant IL-17D (from E. coli)

Supplier: Peprotech

AF-200-27-5UG AF-200-27-25UG AF-200-27-100UG AF-200-27-250UG AF-200-27-500UG AF-200-27-1MG
75999-726EA 266.99 CAD
75999-726 75999-728 75999-730 75999-732 75999-734 75999-736
Human Recombinant IL-17D (from E. coli)
Proteins and Peptides

  • Animal-free


IL-17D is a disulfide-linked homodimer of two 185 amino acid polypeptide chains. It belongs to the IL-17 family of structurally-related cytokines that share a highly conserved C-terminal region, but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17D has the ability to stimulate the production of IL-6, IL-8 and GM-CSF, and inhibits hemopoiesis of myeloid progenitor cells in colony-forming assays. Recombinant Human IL-17D is a 40.5 kDa disulfide-linked homodimer of two 185 amino acid polypeptide chains.

Order Now

SPECIFICATIONS

Protein/peptide name IL-17D
Environmentally Preferable
Protein synonyms Interleukin-17D
Protein/peptide type Recombinant
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID 53342 Q8TAD2
Sequence APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP
Protease-free Y
Animal-free Y
Endotoxin level Low
Endotoxin content >0.1 ng/μg
Biological activity Biological Activity tests can vary. Please refer to the Product CoA for information.
Purity 0.98
Form Lyophilized
Formulation Formulation can vary. Please refer to product CoA for information.
Molecular weight 40.5 kDa
Reconstitution instructions Reconstitution instructions can vary. Please refer to the product CoA for info.
Storage conditions Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR