Human Recombinant FGF-19 (from E. coli)
Supplier: Peprotech
AF-100-32-100UG
AF-100-32-250UG
AF-100-32-500UG
76081-530EA
1845.61
CAD
76081-530
76081-532
76081-534
Human Recombinant FGF-19 (from E. coli)
Proteins and Peptides
- Animal-free
The FGF family plays central roles during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-19, a member of the FGF family, is a high-affinity heparin-dependent ligand for FGFR4. FGF-19 is expressed during brain development and embryogenesis. Recombinant Human FGF-19 is a 21.8 kDa protein containing 195 amino acid residues.