Water USP, Ph. Eur., JP, purified

Supplier: Ricca Chemical

R9189000-20F R9189000-20E1
RCR9189000-20FEA 312.84 CAD
RCR9189000-20F RCR918900020E1
Water USP, Ph. Eur., JP, purified
Water
Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Acidity To Pass Test
Alkalinity To Pass Test
Ammonia (NH₃) max 0.05 ppm
Appearance Colorless liquid
Calcium (Ca) max 0.5 ppm
Carbon Dioxide (CO₂) To Pass Test
Chloride (Cl) max 0.1 ppm
Conductivity at 25°C (Method: SQCP031) max 1 µS/cm
Endotoxins (Chromogenic determination) max 0.25 EU/mL
Heavy Metals (as Pb) max 0.1 ppm
Magnesium (Mg) max 0.5 ppm
Microbial Count (at time of manufacture) max 100 CFU/mL
Nitrate (NO₃) max 1 ppb
Nitrogen (as Nitrite) (NO₂-N) max 1 ppb
Organic Carbon (TOC) max 0.5 ppm
Oxidizable Substances (Permanganate Retention) To Pass Test
pH at 25°C (Method: SQCP027) 5.0-7.0
Residue after evaporation (Non-Volatile Matter) max 0.001 %
Sulfate (SO₄) max 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR