Water ASTM Type IV
Supplier: Ricca Chemical
R9151400-1A
R9151400-20F
R9151400-55E
RCR9151400-1AEA
129.56
CAD
RCR9151400-1A
RCR9151400-20F
RCR9151400-55E
Water ASTM Type IV
Water
Formula:
H₂O Boiling Pt: 100 °C Melting Pt: 0 °C Density: 0.997 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Specification Test Results
Appearance | Colorless liquid |
Conductivity at 25°C | max 5.0 µS/cm |
pH at 25°C | 5.0-8.0 |
Sodium (Na) | max 50 ppb |
Chloride (Cl) | max 50 ppb |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...