VWR® Mini Electrophoresis Systems
Supplier: VWR International
E1103-VWR-E E1103-VWR
76196-448EA 1432.93 CAD
76196-448 76196-486
VWR® Mini Electrophoresis Systems
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
VWR® Mini electrophoresis system is an ideal tool for quick and efficient nucleic acid separations.
  • Complete and convenient system
  • Power supply connects to the gel box, no wires are needed
This system is used for quick nucleic acid separations for a range of applications including evaluation of PCR results. The power supply has 3 voltage choices, connects to the gel tank so no wire leads are needed, and the running tank is compact and does not require a large amount of buffer.
Delivery information: Includes a power supply, gel tank, and gel casting stands, trays and combs, two gel casting sets with combs to make 10.5×6 cm and 5×6 cm gels.