Equal Volume Density Cube Sets
Supplier: United Scientific Supplies
DCSET4 DCSET10
470014-710EA 30 CAD
470014-710 470303-936
Equal Volume Density Cube Sets
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
High quality density cubes for the classroom.
  • Ideal for density experiments and to explore the physical properties of different materials
  • Compare masses for equal volumes
  • Uniform, reliable dimensions
  • Activity manual/reference guide included
Compare the masses and densities of these uniform 1" cubes. Explore the principles of bouyancy, and test the physical properties of metals and non-metals. Each set includes a reference table showing density values of individual specimens.
Ordering information: Set of 4 includes acrylic, pine, nylon, and aluminum, packaged in a poly bag.
Set of 10 includes acrylic, oak, pine, rosewood, steel, aluminum, copper, brass, nylon, and PVC, packaged in a wooden storage box.
Set of 12 includes acrylic, oak, pine, poplar, steel, aluminum, copper, brass, nylon, PVC, lignum vitae, and polypropylene, packaged in a plastic box.