Electric Calorimeter
Supplier: United Scientific Supplies
Ward's®
CLRE02
470304-020EA 48.15 CAD
470304-020
Electric Calorimeter
Calorimeters
A Standard and Electric Calorimeter in one.
  • Electrical requirements are 6 V DC, up to 2A
This device can be used as a classic standard calorimeter or as an electric calorimeter. Made of aluminum and plastic, the calorimeter has a diameter of 10 cm, height of 11 cm, liquid capacity of 175 ml, and a heat capacity of 25 J/°C.