Sulfuric acid 10% (v/v)

Supplier: Ricca Chemical
Danger

R8150000-2.5D
RCR81500002.5DEA 189.6 CAD
RCR81500002.5D
Sulfuric acid 10% (v/v)
Sulfuric acid
Formula: H₂SO₄
Boiling Pt: ∼100 °C
Melting Pt: ∼0 °C
Density: 1.107 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 9.97-10.03% v/v


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR