Ragnell Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-3335 15-3336
76193-044EA 505.68 CAD
76193-044 76193-046
Ragnell Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut Sklarhone™ Ragnell Scissors, also known as Kilner/Ragnell scissors, are finger ring scissors that combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone™ scissors minimize tearing of tissue so cuts and edges are smooth and clean. The long shanks and short jaws of these scissors allow the user greater control with less effort during dissection. This combined with flat blades and blunt tips often make Ragnell scissors the dissection instrument of choice during longer procedures.