Mayo Dissecting Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-3434 15-3435 15-3429 15-3430 15-3427
76191-530EA 743.26 CAD
76191-530 76191-532 76191-594 76191-888 76193-070
Mayo Dissecting Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut Sklarhone™ Mayo Dissecting Scissors are a variation of standard Mayo Scissors that feature one knife-like razor edge and one micro-serrated scissors edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design and the micro-serrations that minimize tearing of tissue and edges so cuts are smooth and clean on all tissue types. Mayo scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference.
Mayo scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference.