Metzenbaum Dissecting Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-3408 15-3411 15-3407
76191-564EA 584.89 CAD
76191-564 76191-566 76191-582
Metzenbaum Dissecting Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut Sklarhone™ Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures or drains. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone™ scissors minimize tearing of tissue so cuts and edges are smooth and clean. Sklarhone™ instruments are identified by dual black finger rings.