Metzenbaum-Lahey Dissecting Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-3303 15-3304 15-3307
76191-822EA 420.04 CAD
76191-822 76191-824 76193-076
Metzenbaum-Lahey Dissecting Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Sklarhone™ Metzenbaum-Lahey Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures or drains. Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors. Sklarhone™ scissors feature one knife-like razor sharp edge and one standard scissor edge. This allows the user to make the sharp incisions of a knife with the cutting ability of scissors. Catching skin and pulls are minimized by the use of Sklarhone™ premium scissors. Sklarhone™ instruments are identified by dual black finger rings.