Dissecting Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-1271 15-1272 15-1274 15-1275 15-1270 15-1273
76191-596EA 424.71 CAD
76191-596 76191-598 76191-600 76191-602 76191-838 76191-840
Dissecting Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
OR-Grade Dissecting Scissors are used primarily for separating tissue or cutting specimen for study. These standard scissors should be used with tissue that is neither too delicate nor too tough, upon which tailored dissecting scissors, such as Mayo or Metzenbaum, would be more appropriate.