Stevens Tenotomy Scissors, OR-Grade, Sklar®
Supplier: Sklar
15-1242 15-1243
76191-544EA 647.48 CAD
76191-544 76191-546
Stevens Tenotomy Scissors, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Supercut Sklarhone™ Stevens Tenotomy Scissors are used for delicate dissection and cutting, commonly in ophthalmologic, neurological, and plastic surgery procedures. Tenotomy scissors have long handles and small, sharp blades with sharp or blunt wedge-shaped tips. The long handles allow the surgeon to have an adequate grip and control while dissecting or cutting in small or narrow areas. Supercut Sklarhone™ scissors combine one knife-like razor edge and one micro-serrated scissor edge. These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone™ scissors minimize tearing of tissue so cuts and edges are smooth and clean. Sklarhone™ instruments are identified by dual black finger rings.