Stille Plaster Cast Shears, OR-Grade, Sklar®
Supplier: Sklar
11-1605 11-1608
76191-494EA 1259.06 CAD
76191-494 76191-706
Stille Plaster Cast Shears, OR-Grade, Sklar®
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Stille Plaster Cast Shears are used for removing plaster casts. The handle is a spring plier type that has a solid serrated blade and a wide fenestrated, cross action blade. This product is designed to use with one hand for smaller casts.