Rotavapor® R-220 Pro Rotary Evaporators, 230 V
Supplier: Buchi
118714C211 118714C242 118714C262 118716C201
76266-276EA 89285.31 CAD
76266-276 76266-298 76266-300 76266-302
Rotavapor® R-220 Pro Rotary Evaporators, 230 V
Evaporators Rotary Evaporators
The Rotavapor® R-220 Pro is designed to fulfill a wide range of distillation applications in the industrial environment. Safety, robustness and easy handling are the key benefits of this system.
  • 230 V model
  • High performance for fastest distillation rates
  • Continuous operation with automatic draining and refilling
  • Economical model to cover essential needs
  • Enhanced lifetime through exclusive use of corrosion-free stainless-steel
  • No risk of product contamination by use of highly chemical resistant materials
  • Seamless 24/7 operation with the robust industrial design
User-friendly design with programmable methods guiding through SOPs, automatic distillation even of challenging foaming samples, and remote monitoring on mobile phone with push notifications. It features convenient flask handling with patented BUCHI flask coupling and unique flask handler. It is maximum safety guaranteed by plastic coated glassware (P+G) and additional protection shields and grids.
Seven glass assemblies available for various uses, including distillation of foaming samples, recrystallization under reflux and height restrictions. System extension is easily possible with a wide range of accessories, such as automatic foam detection, temperature monitoring, coolant flow control.
The 'Essential' version is equipped with a vertical condenser and one receiving flask with a volume of 10 L. This configuration covers all the basic distillation needs. It is not recommended for bumping or foaming samples and also distillations under reflux
are not possible.
Delivery information: Supplied with evaporating flask, all needed tubing's (vacuum and cooling), seal removing tool and operation manual.
Caution: 230 V models.