Vikan® Round Scoops, Remco Products
Supplier: Remco Products
56814 56815 56816 56817 56818 56822 56823 56824 56825 56826 56811 56812 56813
76254-422EA 23.18 CAD
76254-422 76254-424 76254-426 76254-428 76254-430 76254-432 76254-434 76254-436 76254-438 76254-440 76254-442 76254-444 76254-446
Vikan® Round Scoops, Remco Products
Scoops
These full color-coded bowl scoops are great for measuring and scooping liquids.
  • Ergonomically designed round bowl
  • Lightweight and durable
  • Easy to clean
  • Autoclavable at 121 °C (250 °F)
These are designed with a pouring spout on two sides to enable both left and right-hand users to pour from the scoop. The one-piece construction eliminates areas where bacteria can hide. Smooth surface makes it extremely easy to clean, and ideal for use in food production areas for moving food ingredients, liquids etc. There is a measuring scale inside the scoop.
Certifications: FDA Compliant.