Synthware Burets, Kemtech America
Supplier: Kemtech America
F264850 F264525 F264825
76264-362EA 498.73 CAD
76264-362 76264-364 76264-366
Synthware Burets, Kemtech America
Burettes
These high quality burets meet ASTM standards.
  • High quality borosilicate glass
Synthware borosilicate buret is Class A rated for superior accuracy.
Certifications: Meets ASTM Specification E438, Type 1, Class A, and "Standard Specification for Glasses in Laboratory Apparatus".