Barnes Suction Tube, OR-Grade, Sklar®
Supplier: Sklar
07-1830 07-1831 07-1832
76191-880EA 274.42 CAD
76191-880 76191-882 76191-884
Barnes Suction Tube, OR-Grade, Sklar®
Cannulae
Barnes Suction Tube is commonly used in ENT and facial plastic surgery procedures to remove excess fluid from the procedure site.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
It features a smooth thumb plate cutoff to aid in control of the suction as well as the tube itself.