Stirring Shafts, Polished, 19 mm, Chemglass
Supplier: Chemglass
CG-2088-01 CG-2088-03 CG-2088-02
80071-888EA 828.28 CAD
80071-888 80071-892 80071-890
Stirring Shafts, Polished, 19 mm, Chemglass
A precision ground and polished stirrer shaft to fit (80071-866) 19 mm PTFE bearings. It is provided with side button for attachment of (80062-374) PTFE stirrer blades.
These shafts have a reduced diameter top section which is ¹/₂" OD.