Claricep™ Bulk Media, Irregular Alumina, Phenomenex
Supplier: Phenomenex
FCA140100-N FCA1401000-N FCA1405000-N FCA140100-B FCA1401000-B FCA1405000-B FCA140100-A FCA1401000-A FCA1405000-A
89214-292EA 527.61 CAD
89214-292 89214-294 89214-296 89214-298 89214-300 89214-302 89214-304 89214-306 89214-308
Claricep™ Bulk Media, Irregular Alumina, Phenomenex
Chromatography Sorbents Flash Chromatography Sorbents
Alkaline aluminum oxide (PH 7 to 7.5), suitable for neutral or alkaline compounds, such as, Polycyclic hydrocarbon, bio- alkaline, amine, fat-soluble vitamins and aldehyde ketone.
Average particle size: 100 – 200 mesh.