Synthware Desiccators
Supplier: Kemtech America
Synthware
B135121 B135115 B135130 B135124 B135118 B135140
76264-310EA 353.27 CAD
76264-310 76264-314 76264-304 76264-312 76264-308 76264-306
Synthware Desiccators
Desiccators Desiccators, Glass
Our desiccators are made from high quality borosilicate glass.
  • Heavy wall borosilicate construction
  • Ceramic plate included
  • Glass lid included
The desiccator is designed for operation under vacuum pressure and is great for desiccation of your samples or for storage of humidity sensitive materials.
Certifications: Meet ASTM standards.
Delivery information: Supplied with inner ceramic plate and glass lid.