Laboratory Trays, United Scientific Supplies
Supplier: United Scientific Supplies
81701-VWR 81702
76263-312PK 559.99 CAD
76263-312 76263-314 470338-980
Laboratory Trays, United Scientific Supplies
Trays/Dishes/Pots Laboratory Trays/Dishes
General use tray for all laboratory needs.
  • Polypropylene
  • Autoclavable
Multipurpose, polypropylene molded trays can be used for sterilizing, drying glassware, porcelain ware, and other laboratory supplies. These steam autoclavable trays have tapered walls designed for nesting.