Samco™ Exact Volume Transfer Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
941NL 967NL 966NL 930NL 785NL 784NL 783NL 191NL 788NL 787NL 786NL
75838-696CS 1154.54 CAD
75838-696 75838-710 75838-712 75838-714 76196-596 76196-598 76196-600 76196-606 76196-608 76196-610 76196-612
Samco™ Exact Volume Transfer Pipettes, Thermo Scientific
Pipettes Pasteur/Transfer Pipettes
The exact volume transfer pipettes are used for dispensing reagents of small samples.
  • Polyethylene
  • Non sterile
  • Ideal for home and POC (Point of Care) testing
Useful in clinical laboratories. Custom design options available to meet your specifications.