Support Stands with Rods, United Scientific Supplies
Supplier: United Scientific Supplies
54101 54102 54103
76263-066EA 59.97 CAD
76263-066 76263-068 76263-302
Support Stands with Rods, United Scientific Supplies
Retort Stands
All-in-one support design.
  • Non-corrosive
  • Steel rods are coated with polypropylene
Molded polystyrene support stands are ideal for use in applications that require non-corrosive worksurfaces. Heavy, weighted base provides stability similar to metal versions. Each comes with a rod that is 30" long and 0.5" in diameter.