Water-Jacketed CO₂ Incubators, Shel Lab, Sheldon
Supplier: Sheldon Manufacturing
SCO6WE SCO2W SCO5W
75870-824EA 16586.29 CAD
75870-824 75870-832 75870-834
Water-Jacketed CO₂ Incubators, Shel Lab, Sheldon
Incubators CO₂ Incubators
SCO water jacketed CO₂ incubators provide the uniform temperatures and highly stable CO₂ environments necessary to prevent shocks to cell samples. Copper gas plumbing and a glass viewing door enhance the SCO water jacket CO₂ incubators with extensive protection against microbiological contamination.
  • Data logging capabilities
  • Autoclavable shelving system
  • Durable stainless steel chamber for easy cleaning
  • Stable temperature environment maintains constant humidity through evaporation at up to 95%
Infrared (IR) CO₂ sensor is extremely quick to recover CO₂More Product Information