Plastic Spatulas, United Scientific Supplies
Supplier: United Scientific Supplies
81301 81302
76263-282PK 20.56 CAD
76263-282 76263-284
Plastic Spatulas, United Scientific Supplies
Spatulas
Molded in polypropylene, spatulas feature one flat end and one spoon end.
  • Polypropylene
  • Autoclavable
This molded spoon is features an ergonomic deigned which make sit easy to hold.