arium® mini plus Ultrapure Laboratory Water Systems, Sartorius (Starter Packs)
Supplier: Sartorius
H2O-MA-SET-US H2O-MA-UV-SET-US
75994-886EA 13266.72 CAD
75994-886 75994-888
arium® mini plus Ultrapure Laboratory Water Systems, Sartorius (Starter Packs)
Water Purification Systems Pure and Ultra Pure Water Systems
The arium® mini plus is directly connected to a feed water tap. The feed water is treated by a special designed pre-treatment cartridge, which consists of a combination of activated carbon and a reverse osmosis module.
  • Simple bag exchange in less than 5 minutes
  • No use of cleaning agents needed
  • Optimized purity, due to closed system, avoiding a permanent biofilm
  • Intuitive menu navigation with high quality glass touch-display, developed through customer survey
  • Small footprint (only 28 cm width)
  • Volume dispense from 50 ml
  • Delivers a flow rate of 1 liter per minute
The arium® mini plus provides ASTM Types 1 and 3 water quality.
Features intuitive menu navigation for optimal user comfort. Besides the unique bagtank, the systems are equipped with a high-resolution, touch-activated color display. Easy-to-understand icons guide user intuitively through the menu. The Favorites function automatically stores the volume last dispensed. This increases efficiency and prevents errors during repeated dispensing of identical volumes.